Assay Details
| Gene Symbol: KBTBD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130995.5 | XP_003131043.2 | 1476 | 102 | ||
| GenBank mRNA | AK400654.1 | - | 1 | 1413 | 102 | |
More Information
Set Membership: |
|

KBTBD7
Genomic Map