Assay Details
| Gene Symbol: AGMAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127622.4 | XP_003127670.1 | 579 | 72 | ||
| XM_005665032.3 | XP_005665089.2 | 558 | 72 | |||
| GenBank mRNA | AK232655.1 | - | 2 - 3 | 561 | 72 | |
More Information
Set Membership: |
|

AGMAT
Genomic Map