Assay Details
| Gene Symbol: AP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EF418603.1 | - | 6 - 7 | 506 | 94 | |
| GU133632.1 | - | 6 - 7 | 506 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AP3| Gene Symbol: AP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EF418603.1 | - | 6 - 7 | 506 | 94 | |
| GU133632.1 | - | 6 - 7 | 506 | 94 | ||
Set Membership: |
|