Assay Details
| Gene Symbol: nudt19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004903.1 | NP_001004903.1 | 2 - 3 | 1010 | 86 | |
| XM_012960826.2 | XP_012816280.1 | 4 - 5 | 1337 | 86 | ||
| XM_012960828.2 | XP_012816282.1 | 4 - 5 | 1485 | 86 | ||
| XM_012960829.2 | XP_012816283.1 | 4 - 5 | 1290 | 86 | ||
| XM_012960830.1 | XP_012816284.1 | 3 - 4 | 1064 | 86 | ||
| XM_012960831.2 | XP_012816285.1 | 3 - 4 | 1050 | 86 | ||
| XM_012960832.2 | XP_012816286.1 | 3 - 4 | 1050 | 86 | ||
| XM_018092992.1 | XP_017948481.1 | 4 - 5 | 1285 | 86 | ||
| GenBank mRNA | BC075322.1 | - | 2 - 3 | 1010 | 86 | |
More Information
Set Membership: |
|

nudt19
Genomic Map