Assay Details
| Gene Symbol: psmb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203662.1 | NP_988993.1 | 1 - 2 | 177 | 66 | |
| GenBank mRNA | BC061284.1 | - | 1 - 2 | 177 | 66 | |
| CR760776.2 | - | 1 - 2 | 182 | 66 | ||
More Information
Set Membership: |
|

psmb1
Genomic Map