Assay Details
| Gene Symbol: llgl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015691.1 | NP_001015691.1 | 7 - 8 | 772 | 60 | |
| XM_012971729.2 | XP_012827183.1 | 7 - 8 | 794 | 60 | ||
| XM_012971730.2 | XP_012827184.1 | 7 - 8 | 736 | 60 | ||
| GenBank mRNA | AY884236.1 | - | 7 - 8 | 772 | 60 | |
| BC135909.1 | - | 7 - 8 | 786 | 60 | ||
More Information
Set Membership: |
|

llgl2
Genomic Map