Assay Details
| Gene Symbol: dynlrb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016395.1 | NP_001016395.1 | 2 - 3 | 91 | 77 | |
| GenBank mRNA | BC157289.1 | - | 2 - 3 | 91 | 77 | |
| CR761164.1 | - | 2 - 3 | 91 | 77 | ||
More Information
Set Membership: |
|

dynlrb2
Genomic Map