Assay Details
| Gene Symbol: atp5f1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030439.1 | NP_001025610.1 | 7 - 8 | 1014 | 93 | |
| GenBank mRNA | BC091067.1 | - | 7 - 8 | 1014 | 93 | |
More Information
Set Membership: |
|

atp5f1a
Genomic Map