Assay Details
| Gene Symbol: rnf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016554.2 | NP_001016554.1 | 6 - 7 | 518 | 88 | |
| XM_012962491.1 | XP_012817945.1 | 7 - 8 | 755 | 88 | ||
| GenBank mRNA | BC135380.1 | - | 7 - 8 | 672 | 88 | |
| BC157295.1 | - | 6 - 7 | 578 | 88 | ||
| CR760712.2 | - | 6 - 7 | 518 | 88 | ||
More Information
Set Membership: |
|

rnf4
Genomic Map