Assay Details
| Gene Symbol: mob3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016616.2 | NP_001016616.1 | 2 - 3 | 548 | 98 | |
| GenBank mRNA | CR760511.2 | - | 2 - 3 | 548 | 98 | |
More Information
Set Membership: |
|

mob3b
Genomic Map