Assay Details
| Gene Symbol: mob3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016616.2 | NP_001016616.1 | 4 - 5 | 646 | 82 | |
| XM_012962535.2 | XP_012817989.1 | 3 - 4 | 973 | 82 | ||
| GenBank mRNA | CR760511.2 | - | 4 - 5 | 646 | 82 | |
More Information
Set Membership: |
|

mob3b
Genomic Map