Assay Details
| Gene Symbol: rpl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016655.2 | NP_001016655.1 | 2 - 3 | 124 | 66 | |
| GenBank mRNA | BC135496.1 | - | 2 - 3 | 144 | 66 | |
| BC159391.1 | - | 2 - 3 | 126 | 66 | ||
| CR760330.2 | - | 2 - 3 | 124 | 66 | ||
More Information
Set Membership: |
|

rpl22
Genomic Map