Assay Details
| Gene Symbol: rps28p9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016950.2 | NP_001016950.1 | 2 - 3 | 91 | 65 | |
| GenBank mRNA | BC080501.1 | - | 111 | 65 | ||
| CR761699.2 | - | 2 - 3 | 91 | 65 | ||
More Information
Set Membership: |
|

rps28p9
Genomic Map