Assay Details
| Gene Symbol: tsr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017135.2 | NP_001017135.1 | 4 - 5 | 607 | 60 | |
| XM_012970449.1 | XP_012825903.1 | 4 - 5 | 655 | 60 | ||
| XM_012970450.2 | XP_012825904.1 | 4 - 5 | 653 | 60 | ||
| GenBank mRNA | BC153688.1 | - | 3 - 4 | 577 | 60 | |
| BC167337.1 | - | 4 - 5 | 596 | 60 | ||
| CR761043.2 | - | 4 - 5 | 607 | 60 | ||
More Information
Set Membership: |
|

tsr3
Genomic Map