Assay Details
| Gene Symbol: dgat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204041.2 | NP_989372.1 | 3 - 4 | 394 | 64 | |
| GenBank mRNA | BC064191.1 | - | 3 - 4 | 391 | 64 | |
| CR855594.2 | - | 3 - 4 | 394 | 64 | ||
More Information
Set Membership: |
|

dgat2
Genomic Map