Assay Details
| Gene Symbol: enpp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017328.4 | NP_001017328.2 | 3 - 4 | 1182 | 64 | |
| GenBank mRNA | BC121205.1 | - | 3 - 4 | 1182 | 64 | |
More Information
Set Membership: |
|

enpp4
Genomic Map