Assay Details
| Gene Symbol: egfl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078726.1 | NP_001072194.1 | 6 - 7 | 718 | 72 | |
| XM_012968591.2 | XP_012824045.1 | 6 - 7 | 1039 | 72 | ||
| XM_012968592.2 | XP_012824046.1 | 6 - 7 | 1262 | 72 | ||
| XM_012968593.2 | XP_012824047.1 | 6 - 7 | 833 | 72 | ||
| XM_012968594.2 | XP_012824048.1 | 5 - 6 | 943 | 72 | ||
| GenBank mRNA | BC118735.1 | - | 6 - 7 | 718 | 72 | |
| BC136170.1 | - | 6 - 7 | 740 | 72 | ||
| CR761323.2 | - | 6 - 7 | 738 | 72 | ||
More Information
Set Membership: |
|

egfl8
Genomic Map