Assay Details
| Gene Symbol: fezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078995.1 | NP_001072463.1 | 3 - 4 | 1119 | 95 | |
| XM_012959245.2 | XP_012814699.1 | 3 - 4 | 2138 | 95 | ||
| GenBank mRNA | BC121869.1 | - | 3 - 4 | 1119 | 95 | |
| CU075365.1 | - | 3 - 4 | 1320 | 95 | ||
More Information
Set Membership: |
|

fezf1
Genomic Map