Assay Details
| Gene Symbol: tmem170a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113923.1 | NP_001107395.1 | 1 - 2 | 377 | 65 | |
| GenBank mRNA | BC157334.1 | - | 1 - 2 | 377 | 65 | |
| BC170989.1 | - | 1 - 2 | 308 | 65 | ||
| BC170991.1 | - | 1 - 2 | 308 | 65 | ||
| CT025241.2 | - | 1 - 2 | 297 | 65 | ||
More Information
Set Membership: |
|

tmem170a
Genomic Map