Assay Details
| Gene Symbol: pea15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126828.1 | NP_001120300.1 | 2 - 3 | 393 | 87 | |
| XM_012969823.1 | XP_012825277.1 | 2 - 3 | 688 | 87 | ||
| GenBank mRNA | BC160637.1 | - | 2 - 3 | 393 | 87 | |
More Information
Set Membership: |
|

pea15
Genomic Map