Assay Details
| Gene Symbol: rgs19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126653.1 | NP_001120125.1 | 538 | 112 | ||
| GenBank mRNA | BC159034.1 | - | 538 | 112 | ||
| CT027949.1 | - | 479 | 112 | |||
More Information
Set Membership: |
|

rgs19
Genomic Map