Assay Details
| Gene Symbol: tsc22d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012967992.1 | XP_012823446.1 | 2 - 3 | 691 | 84 | |
| XM_012967993.1 | XP_012823447.1 | 2 - 3 | 579 | 84 | ||
| GenBank mRNA | BC161218.1 | - | 2 - 3 | 582 | 84 | |
More Information
Set Membership: |
|

tsc22d3
Genomic Map