Assay Details
| Gene Symbol: LOC100283074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155976.1 | NP_001149448.1 | 767 | 54 | ||
| GenBank mRNA | BT054999.1 | - | 719 | 54 | ||
| EU963367.1 | - | 767 | 54 | |||
More Information
Set Membership: |
|

LOC100283074
Genomic Map