Assay Details
| Gene Symbol: cl3929_1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153189.1 | NP_001146661.1 | 1577 | 103 | ||
| GenBank mRNA | BT055877.1 | - | 1577 | 103 | ||
More Information
Set Membership: |
|

cl3929_1
Genomic Map