Assay Details
| Gene Symbol: BE518809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001151680.1 | NP_001145152.1 | 328 | 94 | ||
| GenBank mRNA | EU973337.1 | - | 328 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BE518809| Gene Symbol: BE518809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001151680.1 | NP_001145152.1 | 328 | 94 | ||
| GenBank mRNA | EU973337.1 | - | 328 | 94 | ||
Set Membership: |
|