Assay Details
| Gene Symbol: LOC100126542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112597.1 | NP_001106067.1 | 2513 | 60 | ||
| GenBank mRNA | EF051578.1 | - | 2513 | 60 | ||
More Information
Set Membership: |
|

LOC100126542
Genomic Map