Assay Details
| Gene Symbol: STY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111806.1 | NP_001105276.1 | 698 | 88 | ||
| GenBank mRNA | AY738149.1 | - | 698 | 88 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STY2| Gene Symbol: STY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111806.1 | NP_001105276.1 | 698 | 88 | ||
| GenBank mRNA | AY738149.1 | - | 698 | 88 | ||
Set Membership: |
|