Assay Details
| Gene Symbol: LOC100191856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137280.1 | NP_001130752.1 | 525 | 101 | ||
| GenBank mRNA | BT040833.1 | - | 442 | 101 | ||
| EU951879.1 | - | 525 | 101 | |||
More Information
Set Membership: |
|

LOC100191856
Genomic Map