Assay Details
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|
||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
| 1000Genome | Applied Biosystems® | HapMap | |||
|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
| C4orf19 - chromosome 4 open reading frame 19 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001104629.1 | 3519 | UTR 3 | NP_001098099.1 | |||
| NM_018302.2 | 3519 | UTR 3 | NP_060772.2 | |||
| XM_011513712.2 | 3519 | UTR 3 | XP_011512014.1 | |||
| XM_011513713.2 | 3519 | UTR 3 | XP_011512015.1 | |||
| XM_017008361.1 | 3519 | UTR 3 | XP_016863850.1 | |||
| RELL1 - RELT like 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001085399.1 | 3519 | Intron | NP_001078868.1 | |||
| NM_001085400.1 | 3519 | Intron | NP_001078869.1 | |||
| XM_017008590.1 | 3519 | Intron | XP_016864079.1 | |||

C4orf19
Genomic Map