Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
2210408I21Rik PubMed Links |
| 2210408I21Rik - RIKEN cDNA 2210408I21 gene | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_001081353.1 | Intron | NP_001074822.1 | ||||
| NM_001145676.1 | Intron | NP_001139148.1 | ||||

2210408I21Rik
Genomic Map