Assay Details
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
4930401G09Rik PubMed Links |
| 4930401G09Rik - RIKEN cDNA 4930401G09 gene | ||||||
|---|---|---|---|---|---|---|
| There are no transcripts associated with this gene. | ||||||
| Msi1 - musashi RNA-binding protein 1 | ||||||
|---|---|---|---|---|---|---|
| Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
| NM_008629.1 | Intron | NP_032655.1 | ||||

4930401G09Rik
Genomic Map