Assay Details
Gene Family ID: | MIPF0000162, mir-367 |
Mature miRNA Sequence: | AAUUGCACUUUAGCAAUGGUGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-367-3p | MIMAT0000719 | |
| Mouse | mmu-miR-367-3p | MIMAT0003181 | |
| Anolis carolinensis | aca-miR-367 | MIMAT0021949 | |
| Columba livia | cli-miR-367-3p | MIMAT0038627 | |
| Dasypus novemcinctus | dno-miR-367-3p | MIMAT0047839 | |
| Guinea pig | cpo-miR-367-3p | MIMAT0047198 | |
| View More Rows | |||
| Horse | eca-miR-367 | MIMAT0012918 | |
| Pan troglodytes | ptr-miR-367 | MIMAT0008118 | |
| Pongo pygmaeus | ppy-miR-367 | MIMAT0015854 | |
| Rabbit | ocu-miR-367-3p | MIMAT0036325 | |
| Rhesus monkey | mml-miR-367 | MIMAT0006292 | |
| Taeniopygia guttata | tgu-miR-367 | MIMAT0025395 | |
Stem-loop Details
Human
Mouse
Anolis carolinensis
Columba livia
Dasypus novemcinctus
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
Taeniopygia guttata
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-367 (v17)
Mouse: mmu-miR-367 (v17)
Mouse: mmu-miR-367 (v17)

Genomic Map