Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUCGUACAGGGUCAUCCACU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gorilla gorilla | ggo-miR-487b | MIMAT0024105 |
Stem-loop Details
Gorilla gorilla
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | ggo-miR-487b |
|---|---|
| miRBase Accession Number: | MI0020652 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AAUCGUACAGGGUCAUCCACU |
| Species: | Gorilla gorilla |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUCGUACAGGGUCAUCCACU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Gorilla gorilla | ggo-miR-487b | MIMAT0024105 |