Assay Details
Gene Family ID: | MIPF0000386, mir-743 |
Mature miRNA Sequence: | UGUUCAGACUGGUGUCCAUCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-743b-5p | MIMAT0004839 |
Stem-loop Details
Mouse
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-743b-5p |
|---|---|
| miRBase Accession Number: | MI0005470 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUUCAGACUGGUGUCCAUCA |
| Species: | Mouse |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000386, mir-743 |
Mature miRNA Sequence: | UGUUCAGACUGGUGUCCAUCA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-743b-5p | MIMAT0004839 |