Assay Details
Amplicon Length: | 82 |
Chromosome Location: | Chr. 3 - 127546025 - 127546025 [-] on 127546025 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-302d-5p | MIMAT0017225 |
| Mouse | mmu-miR-302d-3p | MIMAT0003377 |
| Mouse | mmu-miR-367-5p | MIMAT0017214 |
| Mouse | mmu-miR-367-3p | MIMAT0003181 |
Stem-loop Details
Mouse

Genomic Map