This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ACTG1 SIRNA Assays ›
Gene Symbol:
Actg1
Gene Name:
actin, gamma, cytoplasmic 1
Gene Aliases:
AL023024, Actg, Actl, E51
Chromosome Location:
Chr.11: 120345689 - 120348491 on Build GRCm38
UniGene ID:
Mm.196173
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 100443

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Actg1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009609.2 NP_033739.1 4 812
GenBank mRNA AF195094.1 - 4 842
AK012251.1 - 4 824
AK076081.1 - 4 821
AK087983.1 - 4 821
AK152202.1 - 4 825
AK153446.1 - 4 823
AK157726.1 - 4 821
AK159867.1 - 4 824
AK162157.1 - 4 818
AK183894.1 - 1 202
AK200871.1 - 1 334
AK208587.1 - 1 200
AK208588.1 - 1 200
AK209715.1 - 1 200
BC003337.1 - 4 807 3257312
BC018571.1 - 4 807
BC021796.1 - 4 809 4218945
BC023248.1 - 4 812 3499390
BC099371.1 - 4 804 30841085
M21495.1 - 4 785
X13055.1 - 3 727

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

sarcomere organization
platelet aggregation
nucleotide binding
structural constituent of cytoskeleton
protein binding
ATP binding
ubiquitin protein ligase binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline