siRNA Details
| Gene Symbol: Dctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016890.4 | NP_058586.3 | 6, 7 | 498 | ||
| GenBank mRNA | AF098508.1 | - | 6, 7 | 477 | ||
| AK002720.1 | - | 6, 7 | 489 | |||
| AK003255.1 | - | 6, 7 | 473 | |||
| AK011840.1 | - | 6, 7 | 495 | |||
| AK159344.1 | - | 6, 7 | 496 | |||
| AK182271.1 | - | 2, 3 | 93 | |||
| BC061120.1 | - | 6, 7 | 479 | 6774742 | ||

Dctn3
Genomic Map