This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ADAM6A SIRNA Assays ›
Gene Symbol:
Adam6a Adam6b
Gene Name:
a disintegrin and metallopeptidase domain 6A
a disintegrin and metallopeptidase domain 6B
Gene Aliases:
Adam6, tMDCIV
4930523C11Rik
Chromosome Location:
Chr.12: 113543908 - 113546414 on Build GRCm38
Chromosome Location:
Chr.12: 113489565 - 113491835 on Build GRCm38
UniGene ID:
Mm.188683
Mm.442216
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID 100942

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Adam6a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174885.3 NP_777479.2 1 274
GenBank mRNA AY158689.1 - 1 274
BC108387.1 - 1 227
BC139059.1 - 1 259 8862081
Gene Symbol: Adam6b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009545.1 NP_001009545.1 1 173
GenBank mRNA AK029742.1 - 1 228
AK076580.1 - 1 228
BC111877.1 - 1 193 40059665

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline