This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ANKRD23 SIRNA Assays ›
Gene Symbol:
Ankrd23
Gene Name:
ankyrin repeat domain 23
Gene Aliases:
1110058D09Rik, 2310041C05Rik, AI853437, Darp, MARP3
Chromosome Location:
Chr.1: 36530188 - 36536883 on Build GRCm38
UniGene ID:
Mm.41421
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 100999

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ankrd23
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153502.3 NP_705722.2 5 531
GenBank mRNA AF492400.1 - 5 531
BC022973.1 - 690
BC023221.1 - 5 508
BC132370.1 - 5 541 40130647
BC138797.1 - 5 541 8861819
BC145421.1 - 5 541 9053979

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
response to muscle stretch
regulation of sarcomere organization
titin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline