siRNA Details
| Gene Symbol: Snrpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011432.2 | NP_035562.1 | 3, 4 | 239 | ||
| GenBank mRNA | AK137213.1 | - | 3, 4 | 222 | ||
| AK168973.1 | - | 3, 4 | 212 | |||
| BC008243.1 | - | 3, 4 | 204 | 2655238 | ||
| BC067386.1 | - | 1955 | ||||
| BC092266.1 | - | 3, 4 | 204 | 4009300 | ||
| U70315.1 | - | 3, 4 | 208 | |||
| X96767.1 | - | 3, 4 | 195 | |||

Snrpc
Genomic Map