siRNA Details
| Gene Symbol: Vmn1r11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053233.2 | NP_444463.2 | 1 | 143 | ||
| GenBank mRNA | BC104398.1 | - | 1 | 143 | 40050459 | |
| BC104399.1 | - | 1 | 143 | 40050460 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r11| Gene Symbol: Vmn1r11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053233.2 | NP_444463.2 | 1 | 143 | ||
| GenBank mRNA | BC104398.1 | - | 1 | 143 | 40050459 | |
| BC104399.1 | - | 1 | 143 | 40050460 | ||