siRNA Details
| Gene Symbol: Olfr368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146374.1 | NP_666486.1 | 1 | 260 | ||
| GenBank mRNA | BC104308.1 | - | 1 | 260 | 40044244 | |
| BC104309.1 | - | 1 | 260 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr368| Gene Symbol: Olfr368 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146374.1 | NP_666486.1 | 1 | 260 | ||
| GenBank mRNA | BC104308.1 | - | 1 | 260 | 40044244 | |
| BC104309.1 | - | 1 | 260 | |||