siRNA Details
| Gene Symbol: Grcc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013535.1 | NP_038563.1 | 2 | 237 | ||
| GenBank mRNA | AK009591.1 | - | 2 | 164 | ||
| BC005683.1 | - | 2 | 216 | 3154555 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grcc10| Gene Symbol: Grcc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013535.1 | NP_038563.1 | 2 | 237 | ||
| GenBank mRNA | AK009591.1 | - | 2 | 164 | ||
| BC005683.1 | - | 2 | 216 | 3154555 | ||