This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TLK2 SIRNA Assays ›
Gene Symbol:
TLK2
Gene Name:
tousled like kinase 2
Gene Aliases:
HsHPK, PKU-ALPHA
Chromosome Location:
Chr.17: 62477481 - 62615481 on Build GRCh38
UniGene ID:
Hs.445078
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 103507
Validated

Purification
Size
Availability Inventoried
Catalog # AM51331
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TLK2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284333.1 NP_001271262.1 12 1251
NM_001284363.1 NP_001271292.1 10 912
NM_006852.3 NP_006843.2 12 1254
XM_011524215.2 XP_011522517.1 10 895
XM_011524216.2 XP_011522518.1 12 1015
XM_011524217.2 XP_011522519.1 12 1202
XM_011524218.2 XP_011522520.1 10 895
XM_011524220.2 XP_011522522.1 9 799
XM_011524221.2 XP_011522523.1 11 913
XM_011524222.2 XP_011522524.1 12 1394
XM_011524223.2 XP_011522525.1 10 1056
XM_011524224.2 XP_011522526.1 10 1114
XM_011524225.2 XP_011522527.1 10 1062
XM_011524226.2 XP_011522528.1 12 1108
XM_011524227.2 XP_011522529.1 11 1162
XM_011524228.2 XP_011522530.1 12 1262
XM_011524229.2 XP_011522531.1 13 1189
XM_011524231.1 XP_011522533.1 11 988
XM_017024044.1 XP_016879533.1 12 1088
XM_017024045.1 XP_016879534.1 11 1196
XM_017024046.1 XP_016879535.1 12 1018
XM_017024047.1 XP_016879536.1 12 1015
XM_017024048.1 XP_016879537.1 11 1410
XM_017024049.1 XP_016879538.1 11 1363
XM_017024050.1 XP_016879539.1 10 961
XM_017024051.1 XP_016879540.1 11 1410
XM_017024052.1 XP_016879541.1 11 1361
XM_017024053.1 XP_016879542.1 11 1033
XM_017024054.1 XP_016879543.1 10 1059
XM_017024055.1 XP_016879544.1 9 963
XM_017024056.1 XP_016879545.1 10 1114
XM_017024057.1 XP_016879546.1 10 928
XM_017024058.1 XP_016879547.1 12 1206
XM_017024059.1 XP_016879548.1 12 1196
XM_017024060.1 XP_016879549.1 11 1113
GenBank mRNA AB004884.1 - 10 890
AF162667.1 - 12 1126
AK300586.1 - 10 912
AX775745.1 - 12 1126
BC044925.1 - 12 1158

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191652, XM_006247576, XM_008768350, XM_006247574, XM_008768348, and others.
This siRNA also targets the following transcripts in Mouse: NM_001294334, NM_001112705, NM_001294330, NM_001294331, NM_001294329, and others.

Matching Taqman Gene Expression Assay:


Hs00851784_g1

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of chromatin assembly or disassembly
protein phosphorylation
cellular response to DNA damage stimulus
cell cycle
chromosome segregation
negative regulation of autophagy
covalent chromatin modification
peptidyl-serine phosphorylation
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
intracellular signal transduction
cellular response to gamma radiation
protein serine/threonine kinase activity
protein binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline