siRNA Details
| Gene Symbol: C1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001733.4 | NP_001724.3 | 7 | 1088 | ||
| XM_017019915.1 | XP_016875404.1 | 7 | 1162 | |||
| GenBank mRNA | AK024951.1 | - | 5 | 2094 | ||
| AK222481.1 | - | 7 | 1082 | |||
| AK222491.1 | - | 7 | 1082 | |||
| AK291313.1 | - | 7 | 1110 | |||
| AK298445.1 | - | 7 | 1103 | |||
| AK303750.1 | - | 7 | 981 | |||
| BC035220.1 | - | 7 | 1088 | 4777940 | ||
| M14058.1 | - | 7 | 1092 | |||
| X04701.1 | - | 7 | 1080 | |||

C1R
Genomic Map