This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PKD2L2 SIRNA Assays ›
Gene Symbol:
PKD2L2
Gene Name:
polycystin 2 like 2, transient receptor potential cation channel
Gene Aliases:
TRPP5
Chromosome Location:
Chr.5: 137889420 - 137942747 on Build GRCh38
UniGene ID:
Hs.716884
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 104638

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PKD2L2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258448.1 NP_001245377.1 12, 13 1655
NM_001258449.1 NP_001245378.1 10, 11 1418
NM_001300921.1 NP_001287850.1 12, 13 1721
NM_014386.3 NP_055201.2 12, 13 1721
XM_011543318.2 XP_011541620.1 11, 12 1635
XM_011543321.1 XP_011541623.1 12, 13 1655
XM_017009344.1 XP_016864833.1 11, 12 1845
XM_017009345.1 XP_016864834.1 11, 12 1515
GenBank mRNA AF118125.1 - 12, 13 1688
AF182034.1 - 12, 13 1691
AK301924.1 - 12, 13 1624
BC044581.1 - 10, 11 1417 4830805

Back To Top

More Information


Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

biological_process
detection of mechanical stimulus
calcium ion transmembrane transport
calcium channel activity
calcium ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline