This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CTSD SIRNA Assays ›
Gene Symbol:
CTSD
Gene Name:
cathepsin D
Gene Aliases:
CLN10, CPSD, HEL-S-130P
Chromosome Location:
Chr.11: 1752752 - 1763992 on Build GRCh38
UniGene ID:
Hs.654447
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 105581
Validated

Purification
Size
Availability Inventoried
Catalog # AM51331
Price 6,301.00
Your Price
Online offer:
6301.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CTSD
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001909.4 NP_001900.1 6 892
GenBank mRNA AK130178.1 - 6 784
BC001574.1 - 2 231
BC016320.2 - 6 804 3506977
BT006910.1 - 6 759
BT020155.1 - 6 759
CR456947.1 - 6 759
EU794598.1 - 6 804
KU178007.1 - 6 759
M11233.1 - 6 810
X05344.1 - 6 761

Back To Top

More Information


Matching Taqman Gene Expression Assay:


Hs00157201_m1

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
autophagy
antigen processing and presentation of exogenous peptide antigen via MHC class II
protein catabolic process
collagen catabolic process
aspartic-type endopeptidase activity
cysteine-type endopeptidase activity
serine-type endopeptidase activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-82hgg:80/100.66.79.225:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0