siRNA Details
| Gene Symbol: CPM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005502.2 | NP_001005502.1 | 2, 3 | 211 | ||
| NM_001874.4 | NP_001865.1 | 2, 3 | 243 | |||
| NM_198320.3 | NP_938079.1 | 2, 3 | 229 | |||
| GenBank mRNA | AF368463.1 | - | 2, 3 | 201 | ||
| AK313180.1 | - | 2, 3 | 201 | |||
| BC022276.1 | - | 2, 3 | 201 | 4768771 | ||
| CB959900.1 | - | 228 | ||||
| CB989009.1 | - | 2, 3 | 299 | |||
| CD109360.1 | - | 2, 3 | 228 | |||
| DA205368.1 | - | 2, 3 | 230 | |||
| DB006895.1 | - | 2, 3 | 214 | |||
| J04970.2 | - | 2, 3 | 243 | |||

CPM
Genomic Map