This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TRIB2 SIRNA Assays ›
Gene Symbol:
TRIB2
Gene Name:
tribbles pseudokinase 2
Gene Aliases:
C5FW, GS3955, TRB2
Chromosome Location:
Chr.2: 12716872 - 12742734 on Build GRCh38
UniGene ID:
Hs.467751
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 1059

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TRIB2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021643.3 NP_067675.1 1, 2 1699
GenBank mRNA AK313237.1 - 1, 2 749
AY245544.1 - 1, 2 1682
BC002637.2 - 1, 2 749 3607549
D87119.1 - 1, 2 1488

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
negative regulation of protein kinase activity
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of MAP kinase activity
negative regulation of interleukin-10 biosynthetic process
negative regulation of fat cell differentiation
nucleotide binding
protein kinase activity
protein kinase inhibitor activity
transcription factor binding
mitogen-activated protein kinase kinase binding
ubiquitin protein ligase binding
ubiquitin-protein transferase regulator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hvhj:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline