This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NECTIN1 SIRNA Assays ›
Gene Symbol:
NECTIN1
Gene Name:
nectin cell adhesion molecule 1
Gene Aliases:
CD111, CLPED1, ED4, HIgR, HV1S, HVEC, OFC7, PRR, PRR1, PVRL1, PVRR, PVRR1, SK-12, nectin-1
Chromosome Location:
Chr.11: 119638098 - 119728725 on Build GRCh38
UniGene ID:
Hs.334846
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 106883

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NECTIN1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002855.4 NP_002846.3 6 1338
GenBank mRNA AF060231.1 - 6 1322
BC104948.1 - 6 1244 8143951
BC113471.1 - 6 1240 8322523
X76400.2 - 6 1157

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

lens morphogenesis in camera-type eye
desmosome organization
iron ion transport
immune response
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
signal transduction
axon guidance
cell recognition
single organismal cell-cell adhesion
adherens junction organization
viral entry into host cell
regulation of synapse assembly
retina development in camera-type eye
enamel mineralization
virus receptor activity
receptor binding
protein binding
coreceptor activity
carbohydrate binding
protein homodimerization activity
virion binding
protein heterodimerization activity
cell adhesion molecule binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xxb7z:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline